Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- VVME2
- Entrez Gene ID
-
100233075
- Gene Name
- malate dehydrogenase
- Gene Aliases
-
GSVIVT01016997001
- Chromosome Location
-
Chr.9: 3252713 - 3259466 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U67426.1 |
- |
15 - 16 |
1618 |
95 |
View Full Product Details