Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYCA1
Entrez Gene ID
100233025
Gene Name
myc anthocyanin regulatory protein
Gene Aliases
-
Chromosome Location
Chr.15_NW_002238116: 1014650 - 1019816 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EF193002.3 - 4 - 5 532 90

View Full Product Details