Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO9
Entrez Gene ID
100259221
Gene Name
MLO-like protein 9
Gene Aliases
-
Chromosome Location
Chr.12: 3675368 - 3680357 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU812239.1 - 2 - 3 175 94

View Full Product Details