Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABP1
Entrez Gene ID
100233074
Gene Name
auxin-binding protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 2246865 - 2252576 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU925809.1 - 2 - 3 190 90

View Full Product Details