Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIPK04
Entrez Gene ID
100253180
Gene Name
CBL-interacting protein kinase 04
Gene Aliases
GSVIVT01021541001
Chromosome Location
Chr.10: 3659135 - 3669313 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901232.1 - 1 - 2 166 186

View Full Product Details