Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIPK09
Entrez Gene ID
100262837
Gene Name
CBL-interacting protein kinase 09
Gene Aliases
GSVIVT00016914001
Chromosome Location
Chr.11: 4710816 - 4745118 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901237.1 - 13 - 14 1247 93

View Full Product Details