Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCC1
Entrez Gene ID
100251415
Gene Name
cation chloride co-transporter 1
Gene Aliases
-
Chromosome Location
Chr.10: 4753246 - 4808028 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA GQ161924.1 - 12 - 13 1793 87
GQ161925.1 - 12 - 13 1793 87

View Full Product Details