Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rprd1a
Entrez Gene ID
448020
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Gene Aliases
rprd1b
Chromosome Location
Chr.10: 30935862 - 30974803 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004796.1 NP_001004796.1 4 - 5 581 83
XM_012971622.2 XP_012827076.1 4 - 5 619 83
GenBank mRNA BC074536.1 - 4 - 5 581 83

View Full Product Details