Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pelo
Entrez Gene ID
448078
Gene Name
pelota homolog
Gene Aliases
cgi-17, pelota, pro1770
Chromosome Location
Chr.9: 7066872 - 7085079 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004823.1 NP_001004823.1 - 1063 69
GenBank mRNA BC074604.1 - - 1063 69
CR848246.1 - 9 - 10 1065 69

View Full Product Details