Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
akr1c8p
Entrez Gene ID
448368
Gene Name
aldo-keto reductase family 1, member C8, pseudogene
Gene Aliases
akr1cl1
Chromosome Location
Chr.3: 1492749 - 1500183 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004955.1 NP_001004955.1 8 - 9 953 77
GenBank mRNA BC075451.1 - 8 - 9 953 77

View Full Product Details