Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trafd1
Entrez Gene ID
448446
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
fln29
Chromosome Location
Chr.1: 155071418 - 155090649 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004989.2 NP_001004989.2 11 - 12 1767 64
NM_001244843.1 NP_001231772.1 11 - 12 1842 64
XM_018092226.1 XP_017947715.1 11 - 12 2038 64
GenBank mRNA BC075550.1 - 11 - 12 1761 64
CR942471.1 - 11 - 12 1833 64
CX960166.2 - 2 - 3 275 64

View Full Product Details