Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trafd1
Entrez Gene ID
448446
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
fln29
Chromosome Location
Chr.1: 155071418 - 155090649 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004989.2 NP_001004989.2 7 - 8 991 76
NM_001244843.1 NP_001231772.1 7 - 8 1066 76
XM_018092226.1 XP_017947715.1 7 - 8 1262 76
GenBank mRNA BC075550.1 - 7 - 8 985 76
CR942471.1 - 7 - 8 1057 76

View Full Product Details