The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004991.1 | NP_001004991.1 | 2 - 3 | 245 | 73 |
XM_012971643.2 | XP_012827097.1 | 1 - 2 | 590 | 73 | |
XM_012971644.2 | XP_012827098.1 | 2 - 3 | 393 | 73 | |
XM_012971645.2 | XP_012827099.1 | 2 - 3 | 382 | 73 | |
XM_012971646.2 | XP_012827100.1 | 2 - 3 | 361 | 73 | |
GenBank mRNA | BC075561.1 | - | 2 - 3 | 245 | 73 |
CR761248.1 | - | 1 - 2 | 397 | 73 |