Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fcf1
Entrez Gene ID
448479
Gene Name
FCF1, rRNA-processing protein
Gene Aliases
C14orf111
Chromosome Location
Chr.8: 60956282 - 60967010 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004999.1 NP_001004999.1 2 - 3 145 97
GenBank mRNA BC075594.1 - 2 - 3 145 97
CT025502.2 - 1 - 2 72 97

View Full Product Details