Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndufaf3
Entrez Gene ID
448535
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases
-
Chromosome Location
Chr.4: 126233499 - 126238263 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005023.1 NP_001005023.1 4 - 5 424 71
XM_012960866.2 XP_012816320.1 4 - 5 460 71
GenBank mRNA BC076703.1 - 4 - 5 424 71
CT027947.2 - 4 - 5 402 71

View Full Product Details