Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
apcdd1
Entrez Gene ID
448576
Gene Name
adenomatosis polyposis coli down-regulated 1
Gene Aliases
drapc1, xPgo
Chromosome Location
Chr.6: 89590812 - 89616591 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005044.1 NP_001005044.1 4 - 5 1289 104
GenBank mRNA BC076909.1 - 4 - 5 1289 104

View Full Product Details