Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pacsin2
Entrez Gene ID
496460
Gene Name
protein kinase C and casein kinase substrate in neurons 2
Gene Aliases
pacsin1, sdpii, x-PACSIN2
Chromosome Location
Chr.Un|NW_016683549: 1494 - 36250 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011050.1 NP_001011050.1 9 - 10 1252 105
GenBank mRNA BC084175.1 - 9 - 10 1252 105
CT030501.1 - 9 - 10 1255 105

View Full Product Details