Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
zp4
Entrez Gene ID
394445
Gene Name
zona pellucida glycoprotein 4
Gene Aliases
ZPB, gp37, xlZPB, zbp, zp1, zp4-a, zp4-b
Chromosome Location
Chr.3: 5445779 - 5455882 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203523.1 NP_988854.1 4 - 5 557 81
GenBank mRNA AY079192.1 - 4 - 5 557 81
CR926385.2 - 4 - 5 546 81

View Full Product Details