The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203659.1 | NP_988990.1 | 2 - 3 | 313 | 94 |
GenBank mRNA | BC061301.1 | - | 2 - 3 | 313 | 94 |