Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spop
Entrez Gene ID
394599
Gene Name
speckle type POZ protein
Gene Aliases
spop-a, spop-b, tef2
Chromosome Location
Chr.10: 9889221 - 9900382 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203672.1 NP_989003.1 9 - 10 1110 76
XM_012971593.2 XP_012827047.1 9 - 10 1344 76
XM_012971594.2 XP_012827048.1 9 - 10 1144 76
XM_012971596.2 XP_012827050.1 9 - 10 1122 76
XM_018097685.1 XP_017953174.1 9 - 10 1170 76
GenBank mRNA BC061316.1 - 9 - 10 1110 76

View Full Product Details