Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trnt1
Entrez Gene ID
394602
Gene Name
tRNA nucleotidyl transferase, CCA-adding, 1
Gene Aliases
cca1, cgi-47, mtcca, xcca
Chromosome Location
Chr.4: 119400266 - 119407454 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012960722.1 XP_012816176.1 3 - 4 357 108
XM_018092956.1 XP_017948445.1 3 - 4 361 108
GenBank mRNA BC061356.1 - 3 - 4 515 108

View Full Product Details