Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
serp1
Entrez Gene ID
394685
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
ramp4
Chromosome Location
Chr.5: 114903565 - 114907253 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203754.1 NP_989085.1 2 - 3 321 66
GenBank mRNA BC062484.1 - 2 - 3 321 66
CR760506.2 - 2 - 3 183 66

View Full Product Details