Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekhj1
Entrez Gene ID
394773
Gene Name
pleckstrin homology domain containing J1
Gene Aliases
gnrpx
Chromosome Location
Chr.1: 83631732 - 83642824 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203835.1 NP_989166.1 3 - 4 413 88
XM_012958309.2 XP_012813763.1 3 - 4 300 88
GenBank mRNA BC063351.1 - 3 - 4 413 88
CR761359.2 - 3 - 4 416 88

View Full Product Details