Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cpt2
Entrez Gene ID
394801
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
cg2107
Chromosome Location
Chr.4: 86950247 - 86960538 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203862.2 NP_989193.1 4 - 5 1678 71
GenBank mRNA BC063210.1 - 4 - 5 1681 71

View Full Product Details