Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
copz1
Entrez Gene ID
394893
Gene Name
coatomer protein complex subunit zeta 1
Gene Aliases
copz, copz2, zeta1-cop
Chromosome Location
Chr.2: 142370446 - 142382345 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203948.1 NP_989279.1 3 - 4 190 88
XM_012956670.2 XP_012812124.1 4 - 5 297 87
XM_012956671.2 XP_012812125.1 3 - 4 258 87
XM_012956672.2 XP_012812126.1 4 - 5 237 87
GenBank mRNA BC064149.1 - 3 - 4 190 88
CR760836.2 - 3 - 4 174 87

View Full Product Details