Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp6v0d2
Entrez Gene ID
394992
Gene Name
ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2
Gene Aliases
-
Chromosome Location
Chr.6: 110524533 - 110541709 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204031.1 NP_989362.1 7 - 8 923 95
GenBank mRNA BC064198.1 - 7 - 8 923 95
CR760271.2 - 7 - 8 923 95

View Full Product Details