Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnn1
Entrez Gene ID
407907
Gene Name
calponin 1, basic, smooth muscle
Gene Aliases
XclpH3, clpH3, cnn2
Chromosome Location
Chr.1: 81168917 - 81179237 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213676.2 NP_998841.1 3 - 4 350 100
GenBank mRNA BC067950.2 - 3 - 4 350 100
CT025165.2 - 3 - 4 327 100

View Full Product Details