Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
echs1
Entrez Gene ID
448019
Gene Name
enoyl-CoA hydratase, short chain, 1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.7: 59096355 - 59152619 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005437.1 NP_001005437.1 7 - 8 822 83
XM_018095333.1 XP_017950822.1 7 - 8 846 83
GenBank mRNA BC074535.1 - 7 - 8 822 83

View Full Product Details