Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppm1j
Entrez Gene ID
448275
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
Gene Aliases
pp2cz, pp2czeta, ppm1h, ppp2cz
Chromosome Location
Chr.2: 74191185 - 74219495 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005797.1 NP_001005797.1 7 - 8 1411 104
GenBank mRNA BC075345.1 - 7 - 8 1411 104

View Full Product Details