Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankra2
Entrez Gene ID
448315
Gene Name
ankyrin repeat family A member 2
Gene Aliases
ankra
Chromosome Location
Chr.1: 166476052 - 166486369 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006691.1 NP_001006692.1 3 - 4 570 104
XM_012959488.2 XP_012814942.1 4 - 5 678 104
XM_012959498.2 XP_012814952.1 4 - 5 1284 104
XM_012959503.2 XP_012814957.1 4 - 5 715 104
XM_012959507.2 XP_012814961.1 5 - 6 1234 104
XM_012959516.2 XP_012814970.1 4 - 5 613 104
GenBank mRNA BC075392.1 - 3 - 4 570 104

View Full Product Details