Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
c12orf4
Entrez Gene ID
493240
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
-
Chromosome Location
Chr.3: 2449852 - 2483554 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007513.1 NP_001007514.1 6 - 7 976 104
XM_012958864.1 XP_012814318.1 6 - 7 1038 104
XM_012958865.1 XP_012814319.1 6 - 7 1035 104
XM_012958867.2 XP_012814321.1 6 - 7 1018 104
XM_012958868.2 XP_012814322.1 6 - 7 1008 104
XM_012958869.1 XP_012814323.1 6 - 7 835 104
XM_012958870.2 XP_012814324.1 6 - 7 852 104
XM_012958871.2 XP_012814325.1 6 - 7 837 104
GenBank mRNA BC079954.1 - 6 - 7 976 104
CT030587.1 - 6 - 7 817 104

View Full Product Details