Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aprt
Entrez Gene ID
493317
Gene Name
adenine phosphoribosyltransferase
Gene Aliases
-
Chromosome Location
Chr.4: 60857496 - 60864065 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007940.1 NP_001007941.1 4 - 5 522 91
XM_018093014.1 XP_017948503.1 4 - 5 390 91
GenBank mRNA BC080448.1 - 4 - 5 522 91
CR761604.2 - 4 - 5 533 91

View Full Product Details