Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
foxo1
Entrez Gene ID
493378
Gene Name
forkhead box O1
Gene Aliases
fkh1, fkhr, foxo1a, xFoxO1
Chromosome Location
Chr.2: 147477412 - 147515842 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008015.1 NP_001008016.1 1 - 2 673 101
GenBank mRNA BC080894.1 - 1 - 2 673 101

View Full Product Details