Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klhl13
Entrez Gene ID
493457
Gene Name
kelch like family member 13
Gene Aliases
bklhd2
Chromosome Location
Chr.8: 45786571 - 45833438 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008094.1 NP_001008095.1 3 - 4 491 96
XM_012968271.2 XP_012823725.1 4 - 5 695 96
XM_018096227.1 XP_017951716.1 3 - 4 546 96
GenBank mRNA BC081294.1 - 3 - 4 491 96
CT025398.2 - 3 - 4 485 96

View Full Product Details