Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nol12
Entrez Gene ID
493530
Gene Name
nucleolar protein 12
Gene Aliases
nop25
Chromosome Location
Chr.4: 112220947 - 112233652 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008167.1 NP_001008168.1 2 - 3 149 76
XM_012960965.2 XP_012816419.1 2 - 3 227 76
XM_012960966.2 XP_012816420.1 2 - 3 238 76
XM_012960967.1 XP_012816421.1 2 - 3 263 76
XM_012960968.2 XP_012816422.1 2 - 3 235 76
GenBank mRNA BC082486.1 - 2 - 3 149 76
CR942728.2 - 2 - 3 150 76

Gene Symbol
LOC101730686
Entrez Gene ID
101730686
Gene Name
nucleolar protein 12-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016687173: 819 - 4025 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004920837.3 XP_004920894.2 2 - 3 270 76
XM_012955444.1 XP_012810898.1 2 - 3 269 76
XM_012955445.2 XP_012810899.1 2 - 3 288 76
XM_018090708.1 XP_017946197.1 2 - 3 280 76

View Full Product Details