Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clmp
Entrez Gene ID
493539
Gene Name
CXADR-like membrane protein
Gene Aliases
acam, asam
Chromosome Location
Chr.7: 54557950 - 54614153 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008176.1 NP_001008177.1 6 - 7 1107 111
XM_012966482.2 XP_012821936.1 7 - 8 1123 111
XM_012966483.2 XP_012821937.1 7 - 8 1119 111
XM_012966484.1 XP_012821938.1 7 - 8 1008 111
GenBank mRNA BC082496.1 - 6 - 7 1107 111

View Full Product Details