Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trim72
Entrez Gene ID
493550
Gene Name
tripartite motif containing 72, E3 ubiquitin protein ligase
Gene Aliases
Mg53
Chromosome Location
Chr.2: 143383796 - 143390136 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008187.1 NP_001008188.1 6 - 7 912 97
GenBank mRNA BC082513.1 - 6 - 7 912 97

View Full Product Details