Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acp5
Entrez Gene ID
493572
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
TRAP-5b
Chromosome Location
Chr.3: 128526876 - 128547543 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008209.1 NP_001008210.1 4 - 5 797 59
XM_012953156.2 XP_012808610.1 4 - 5 773 59
GenBank mRNA BC081357.1 - 4 - 5 797 59
CR942420.2 - 4 - 5 770 59

View Full Product Details