Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdr83
Entrez Gene ID
496482
Gene Name
WD repeat domain 83
Gene Aliases
morg1
Chromosome Location
Chr.3: 129336628 - 129351239 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011071.1 NP_001011071.1 4 - 5 475 66
XM_012953486.2 XP_012808940.1 4 - 5 456 66
GenBank mRNA BC084454.1 - 4 - 5 475 66
CR761781.2 - 3 - 4 383 66

View Full Product Details