Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rdh10
Entrez Gene ID
496504
Gene Name
retinol dehydrogenase 10
Gene Aliases
SDR16C4, XRDH10, XRDH10a, XRDH10b, rdh10-a, rdh10-b
Chromosome Location
Chr.6: 106242680 - 106257403 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011091.1 NP_001011091.1 2 - 3 863 94
GenBank mRNA BC084483.1 - 2 - 3 863 94
CT030281.1 - 2 - 3 858 94

View Full Product Details