Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snx10
Entrez Gene ID
496527
Gene Name
sorting nexin 10
Gene Aliases
-
Chromosome Location
Chr.6: 33392962 - 33414564 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011114.2 NP_001011114.1 3 - 4 191 101
XM_012964580.2 XP_012820034.1 3 - 4 271 101
XM_012964581.2 XP_012820035.1 3 - 4 283 101
XM_012964582.2 XP_012820036.1 2 - 3 274 101
GenBank mRNA BC084512.1 - 3 - 4 192 101
CR761975.1 - 3 - 4 184 101

View Full Product Details