Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps16
Entrez Gene ID
496563
Gene Name
ribosomal protein S16
Gene Aliases
-
Chromosome Location
Chr.3: 42690100 - 42695259 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011146.1 NP_001011146.1 5 - 6 332 103
GenBank mRNA BC084534.1 - 5 - 6 332 103
CR942750.2 - 4 - 5 302 103

View Full Product Details