Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
iyd
Entrez Gene ID
496718
Gene Name
iodotyrosine deiodinase
Gene Aliases
-
Chromosome Location
Chr.5: 54672274 - 54680602 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011267.1 NP_001011267.1 2 - 3 407 98
GenBank mRNA BC087975.1 - 2 - 3 407 98

View Full Product Details