Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
iyd
Entrez Gene ID
496718
Gene Name
iodotyrosine deiodinase
Gene Aliases
-
Chromosome Location
Chr.5: 54672274 - 54680602 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011267.1 NP_001011267.1 4 - 5 720 89
GenBank mRNA BC087975.1 - 4 - 5 720 89

View Full Product Details