Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpm
Entrez Gene ID
496721
Gene Name
carboxypeptidase M
Gene Aliases
cpd
Chromosome Location
Chr.3: 49515986 - 49554740 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011270.1 NP_001011270.1 4 - 5 452 82
XM_012958933.2 XP_012814387.1 5 - 6 858 82
XM_012958934.2 XP_012814388.1 4 - 5 665 82
XM_012958935.2 XP_012814389.1 4 - 5 1392 82
XM_012958937.2 XP_012814391.1 3 - 4 309 82
XM_018092056.1 XP_017947545.1 5 - 6 854 82
GenBank mRNA BC087979.1 - 4 - 5 452 82

View Full Product Details