Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a4
Entrez Gene ID
394504
Gene Name
solute carrier family 25 member 4
Gene Aliases
ant, ant1, peo2, peo3
Chromosome Location
Chr.1: 35375273 - 35384398 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203578.1 NP_988909.1 2 - 3 656 86
GenBank mRNA BC061600.1 - 2 - 3 656 86
CR761556.2 - 2 - 3 676 86

View Full Product Details