Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrpl44
Entrez Gene ID
496841
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
-
Chromosome Location
Chr.5: 125194838 - 125201299 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011374.1 NP_001011374.1 2 - 3 632 61
GenBank mRNA BC088549.1 - 2 - 3 632 61

View Full Product Details