Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
qdpr
Entrez Gene ID
496990
Gene Name
quinoid dihydropteridine reductase
Gene Aliases
-
Chromosome Location
Chr.1: 16476928 - 16487138 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011492.1 NP_001011492.1 1 - 2 111 80
GenBank mRNA BC088878.1 - 1 - 2 111 80

View Full Product Details