Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cela1
Entrez Gene ID
496993
Gene Name
chymotrypsin like elastase family member 1
Gene Aliases
ela1
Chromosome Location
Chr.2: 139150870 - 139157859 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011493.1 NP_001011493.1 3 - 4 241 75
GenBank mRNA BC088882.1 - 3 - 4 241 75

View Full Product Details