Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lyg2
Entrez Gene ID
548456
Gene Name
lysozyme G-like 2
Gene Aliases
-
Chromosome Location
Chr.2: 109180323 - 109193968 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015739.1 NP_001015739.1 3 - 4 371 106
GenBank mRNA BC089674.1 - 3 - 4 371 106

View Full Product Details